Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ki67/MKI67 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF7617
This item is not returnable.
View return policy
Description
Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Human samples. It is validated for Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.Specifications
Ki67/MKI67 | |
Polyclonal | |
Unconjugated | |
P46013 | |
MKI67 | |
E. coli-derived recombinant human Ki-67/MKI67 Asn3120-Ile3256 Accession # P46013 | |
100 μg | |
Primary | |
Detects human Ki-67/MKI67 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Ki-67/MKI67 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay | |
LYOPH | |
Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126 | |
Sheep | |
Affinity Purified | |
RUO | |
4288 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction