Learn More
Description
Specifications
Specifications
| Quantity | 0.1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Membrane Protein - Lupus: Kidney |
| Cellular Fraction | Membrane |
| Tissue Condition | Lupus |
| Cross Reactivity | Human |
| Host Species | Human |
| Product Type | Human Kidney Membrane Tissue Lysate (Adult Lupus) |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Life Stage | Adult |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
