Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR/CD158 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1848
This item is not returnable.
View return policy
Description
KIR/CD158 Monoclonal specifically detects KIR/CD158 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
KIR/CD158 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
CD158, CD158 antigen-like family member A, CD158a, CD158a antigen, CD158Ankat1, CD158b1, CD158b2, CD158d, CD158e2, CD158f, CD158i, CD158k, cl-42, CL-43, CL-5, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, KIR2DL5A, KIR-G1, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, nkat10, NKAT1KIR-K64, NKAT2, NKAT4A, NKAT4B, NKAT6, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3802 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, CyTOF | |
180704 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIR2DL1 | |
BaF3 mouse pro-B cell line transfected with human KIR2DL3 | |
500 μg | |
Primary | |
Detects human KIR/CD158. Stains cells transfected with KIR2DL2, 2DL3, 2DS2, or 2DS4. It does not stain cells transfected with KIR2DL1, 2DL4, 3DL1, or 3DL2. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction