Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR2DL1/KIR2DS5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
$200.50 - $2020.00
Specifications
Antigen | KIR2DL1/KIR2DS5 |
---|---|
Clone | 143211 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported |
Applications | Flow Cytometry, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1844SP
![]() |
R&D Systems
MAB1844SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1844100
![]() |
R&D Systems
MAB1844100 |
100 μg |
Each for $538.00
|
|
|||||
MAB1844500
![]() |
R&D Systems
MAB1844500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry, CyTOF-reported.Specifications
KIR2DL1/KIR2DS5 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD158a, CD158g, NKAT9 | |
KIR2DL1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
143211 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported | |
Monoclonal | |
Mouse | |
Human | |
P43626 | |
3802 | |
BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title