Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR2DL3/CD158b2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2014N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2014N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2014N100 Supplier R&D Systems Supplier No. FAB2014N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR2DL3/CD158b2 Monoclonal specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DL3/CD158b2
Applications Flow Cytometry
Classification Monoclonal
Clone 180701
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P43628
Gene Alias CD158 antigen-like family member B2, CD158b2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, nkat2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
Gene Symbols KIR2DL3
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2 Accession # P43628
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3804
Test Specificity Detects human KIR2DL3/CD158b2. Stains BaF3 cells transfected with human KIR2DL3 in flow cytometry. Does not stain BaF3 cells transfected with KIR2DL1, 2DL2, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.