Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR2DL3/CD158b2 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 9 publications

$174.60 - $385.50

Specifications

Antigen KIR2DL3/CD158b2
Clone 180701
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB2014P025
GREENER_CHOICE
View Documents
R&D Systems
FAB2014P025
25 Tests
Each for $174.60
Only null left
Add to Cart
 
FAB2014P100
GREENER_CHOICE
View Documents
R&D Systems
FAB2014P100
100 Tests
Each for $385.50
Only null left
Add to Cart
 
Description

Description

KIR2DL3/CD158b2 Monoclonal specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

KIR2DL3/CD158b2
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD158 antigen-like family member B2, CD158b2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, nkat2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
KIR2DL3
IgG2a
Protein A or G purified from hybridoma culture supernatant
Detects human KIR2DL3/CD158b2. Stains BaF3 cells transfected with human KIR2DL3 in flow cytometry. Does not stain BaF3 cells transfected with KIR2DL1, 2DL2, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
180701
Flow Cytometry
R-PE
Mouse
Human
P43628
3804
BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2 Accession # P43628
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.