Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR2DS1/CD158h Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8887N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8887N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8887N100 Supplier R&D Systems Supplier No. FAB8887N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIR2DS1/CD158h Monoclonal specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DS1/CD158h
Applications Flow Cytometry
Classification Monoclonal
Clone 1127B
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q14954
Gene Alias killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, EB6ActI, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1
Host Species Rabbit
Immunogen HEK293 human embryonic kidney cell line transfected with human KIR2DS1/CD158h Met1-His245 Accession # Q14954
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3806
Test Specificity Detects human KIR2DS1/CD158h in flow cytometry. Clone 1127B recognizes KIR2DS1 and some alleles of KIR2DL1, both members of the killer cell immunoglobulin-like receptor (KIR) family. Because Clone 1127B displays partial cross-reactivity with KIR2DL1, co-staining with Mouse Anti-Human KIR2DL1 Clone 143211 (Catalog # FAB1844F) is recommended.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.