Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR2DS5/CD158g Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8920R025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8920R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8920R025 Supplier R&D Systems Supplier No. FAB8920R025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIR2DS5/CD158g Monoclonal specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DS5/CD158g
Applications Flow Cytometry
Classification Monoclonal
Clone 1165A
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q14953
Gene Alias killer cell immunoglobulin-like receptor 2DS5, CD158g, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9
Gene Symbols KIR2DS5
Host Species Rabbit
Immunogen NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g His22-His245 Accession # Q14953
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3810
Test Specificity Detects human KIR2DS5/CD158g in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.