Learn More
Human KIR2DS5/CD158g Alexa Fluor 700-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | KIR2DS5/CD158g |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1165A |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q14953 |
| Gene Alias | killer cell immunoglobulin-like receptor 2DS5, CD158g, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9 |
| Host Species | Rabbit |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.