Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR2DS5/CD158g Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $467.50
Specifications
Antigen | KIR2DS5/CD158g |
---|---|
Clone | 1165A |
Dilution | Flow Cytometry 1:1000 dilution |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
KIR2DS5/CD158g Monoclonal specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DS5/CD158g | |
Flow Cytometry 1:1000 dilution | |
Monoclonal | |
Purified | |
RUO | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
killer cell immunoglobulin-like receptor 2DS5, CD158g, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9 | |
KIR2DS5 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Detects human KIR2DS5/CD158g in direct ELISAs. |
1165A | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
Q14953 | |
3810 | |
NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g His22-His245 Accession # Q14953 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title