Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR3DL1 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA12251V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA12251V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA12251V100 Supplier R&D Systems Supplier No. FAB12251V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR3DL1 Monoclonal specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR3DL1
Applications Flow Cytometry
Classification Monoclonal
Clone 177407
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P43629
Gene Alias AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11
Gene Symbols KIR3DL1
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL1 Accession # P43629
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3811
Test Specificity Detects human KIR3DL1 on transfected BaF/3 cells. When tested against a panel of transfectants, no cross-reactivity with KIR2DL1, 2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL2, or 3DS1 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.