Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR3DL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB12251SP
This item is not returnable.
View return policy
Description
KIR3DL1 Monoclonal specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
KIR3DL1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P43629 | |
KIR3DL1 | |
BaF3 mouse pro-B cell line transfected with human KIR3DL1 Accession # P43629 | |
25 μg | |
Primary | |
Detects human KIR3DL1 on transfected BaF/3 cells. When tested against a panel of transfectants, no cross-reactivity with KIR2DL1, 2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL2, or 3DS1 is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
177407 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3811 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction