Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR3DL2/CD158k Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2878U100UG
This item is not returnable.
View return policy
Description
KIR3DL2/CD158k Monoclonal specifically detects KIR3DL2/CD158k in Human samples. It is validated for Flow Cytometry.Specifications
KIR3DL2/CD158k | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P43630 | |
KIR3DL2 | |
BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k Accession # P43630 | |
RUO | |
3812 | |
Human | |
IgG1 |
Flow Cytometry | |
539304 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
killer cell immunoglobulin-like receptor 3DL2, CD158K, CL-5, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4A, NKAT4B, p140 | |
Mouse | |
100 μg | |
Primary | |
Detects human KIR3DL2/CD158k in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction