Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR3DL2/CD158k Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$139.50 - $432.50
Specifications
Antigen | KIR3DL2/CD158k |
---|---|
Clone | 539304 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2878N025
![]() |
R&D Systems
FAB2878N025 |
25 Tests |
Each for $139.50
|
|
|||||
FAB2878N100
![]() |
R&D Systems
FAB2878N100 |
100 Tests |
Each for $432.50
|
|
|||||
Description
KIR3DL2/CD158k Monoclonal specifically detects KIR3DL2/CD158k in Human samples. It is validated for Flow Cytometry.Specifications
KIR3DL2/CD158k | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
killer cell immunoglobulin-like receptor 3DL2, CD158K, CL-5, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4A, NKAT4B, p140 | |
KIR3DL2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human KIR3DL2/CD158k in direct ELISAs. |
539304 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
Human | |
P43630 | |
3812 | |
BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k Accession # P43630 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title