Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR3DL2/CD158k APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

$162.90 - $491.00

Specifications

Antigen KIR3DL2/CD158k
Clone 539304
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB2878A025
GREENER_CHOICE
View Documents
R&D Systems
FAB2878A025
25 Tests
Each for $162.90
Only null left
Add to Cart
 
FAB2878A100
GREENER_CHOICE
View Documents
R&D Systems
FAB2878A100
100 Tests
Each for $491.00
Only null left
Add to Cart
 
Description

Description

KIR3DL2/CD158k Monoclonal specifically detects KIR3DL2/CD158k in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

KIR3DL2/CD158k
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
killer cell immunoglobulin-like receptor 3DL2, CD158K, CL-5, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4A, NKAT4B, p140
KIR3DL2
IgG1
Protein A or G purified from hybridoma culture supernatant
Detects human KIR3DL2/CD158k in direct ELISAs.
539304
Flow Cytometry
APC
Mouse
Human
P43630
3812
BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k Accession # P43630
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.