Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR3DL3/CD158z Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$200.16 - $532.27

Specifications

Antigen KIR3DL3/CD158z
Clone 1136B
Dilution Flow Cytometry 0.25 ug/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB8919SP
GREENER_CHOICE
View Documents
R&D Systems
MAB8919SP
25 μg
Each for $200.16
Only null left
Add to Cart
 
MAB8919
GREENER_CHOICE
View Documents
R&D Systems
MAB8919
100 μg
Each for $532.27
Only null left
Add to Cart
 
Description

Description

KIR3DL3/CD158z Monoclonal specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

KIR3DL3/CD158z
Flow Cytometry 0.25 ug/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
115653
Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z Met1-Leu322 Accession # Q8N743
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human KIR3DL3/CD158z in direct ELISAs.
1136B
Flow Cytometry
Unconjugated
Rabbit
Human
Q8N743
KIR3DL3
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.