Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Kirrel2/NEPH3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2564
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2564 100 μg
AF2564SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2564 Supplier R&D Systems Supplier No. AF2564
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Kirrel2/NEPH3 Polyclonal specifically detects Kirrel2/NEPH3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Kirrel2/NEPH3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q6UWL6
Gene Alias DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2
Gene Symbols KIRREL2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kirrel2/NEPH3 Gly21-Arg503 Accession # Q6UWL6
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84063
Test Specificity Detects human Kirrel2/NEPH3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Kirrel2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.