Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human KIRREL3 ELISA Kit

Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Description
Human KIRREL3 quantitates human KIRREL3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human KIRREL3.
The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. The protein encoded by this gene is a synaptic cell adhesion molecule with multiple extracellular immunoglobulin-like domains and a cytoplasmic PDZ domain-binding motif. Mutations in this gene are associated with several neurological and cognitive disorders. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications
Specifications
Accession Number | 1500010O20Rik; 2900036G11Rik; KIAA1867; kin of IRRE like 3 (Drosophila); Kin of irregular chiasm-like protein 3; kin of IRRE-like protein 3; KIRRE; KIRREL3; membrane protein mKirre; mKIAA1867; mKirre; MRD4; NEPH2; nephrin-like 2; nephrin-like protein 2; PRO4502; Processed kin of IRRE-like protein 3; SST4; UNQ5923/PRO4502/PRO19814; x kin of IRRE like 3 |
Assay Range | 0.21 to 50 ng/mL |
Assay Sensitivity | 0.21 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Sample Type | Plasma, Serum, Supernatant |
For Use With (Equipment) | Colorimetric Microplate Reader |
Gene ID (Entrez) | 84623 |
Gene Symbol | KIRREL3 |
Interassay CV | <12% |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction