Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KLF2 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5466V100UG
This item is not returnable.
View return policy
Description
KLF2 Monoclonal specifically detects KLF2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| KLF2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Y5W3 | |
| KLF2 | |
| E. coli-derived recombinant human KLF2 Pro71-Pro168 (predicted) Accession # Q9Y5W3 | |
| RUO | |
| 10365 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 665333 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human KLF2 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) KLF17 and no cross-reactivity with rhKLF1, 4, 5, 6, 10, 12, 17, recombinant mouse KLF4 or 15 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction