Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KLF2 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5466T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5466T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5466T100 Supplier R&D Systems Supplier No. IC5466T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLF2 Monoclonal specifically detects KLF2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen KLF2
Applications Flow Cytometry
Classification Monoclonal
Clone 665333
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9Y5W3
Gene Alias Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor
Gene Symbols KLF2
Host Species Mouse
Immunogen E. coli-derived recombinant human KLF2 Pro71-Pro168 (predicted) Accession # Q9Y5W3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10365
Test Specificity Detects human KLF2 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) KLF17 and no cross-reactivity with rhKLF1, 4, 5, 6, 10, 12, 17, recombinant mouse KLF4 or 15 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.