Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KLF2 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5466P025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5466P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5466P025 Supplier R&D Systems Supplier No. IC5466P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLF2 Monoclonal specifically detects KLF2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen KLF2
Applications Flow Cytometry
Classification Monoclonal
Clone 665333
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9Y5W3
Gene Alias Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor
Gene Symbols KLF2
Host Species Mouse
Immunogen E. coli-derived recombinant human KLF2 Pro71-Pro168 (predicted) Accession # Q9Y5W3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10365
Test Specificity Detects human KLF2 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) KLF17 and no cross-reactivity with rhKLF1, 4, 5, 6, 10, 12, recombinant mouse KLF4 or 15 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.