Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Klotho Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5334SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5334SP 25 μg
MAB5334100 100 μg
MAB5334500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB5334SP Supplier R&D Systems Supplier No. MAB5334SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Klotho Monoclonal specifically detects Klotho in Human samples. It is validated for Neutralization.

Specifications

Antigen Klotho
Applications Neutralization
Classification Monoclonal
Clone 775340
Conjugate Unconjugated
Dilution Neutralization 0.15-0.6 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UEF7
Gene Alias EC 3.2.1, EC 3.2.1.31, KL, klotho
Gene Symbols KL
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Klotho Glu34-Ser981 Accession # Q9UEF7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9365
Test Specificity Detects human Klotho in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) Klotho (aa 35-982) is observed and no cross-reactivity with rmKlotho (aa 23-550), recombinant human (rh) Klotho beta (aa 53-997), or rhCytosolic beta-Glucosidase/GBA3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.