Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Klotho beta Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA58891T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA58891T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA58891T100 Supplier R&D Systems Supplier No. FAB58891T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Klotho beta Monoclonal specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Klotho beta
Applications Flow Cytometry
Classification Monoclonal
Clone 1025C
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q86Z14
Gene Alias beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived recombinant human Klotho beta with a C-terminal 10 His tag. Phe53-Leu997 Accession # Q86Z14
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152831
Test Specificity Detects human Klotho beta in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.