Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Klotho beta Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB58891S100UG
This item is not returnable.
View return policy
Description
Klotho beta Monoclonal specifically detects Klotho beta in Human samples. It is validated for Flow Cytometry.Specifications
Klotho beta | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
Mouse myeloma cell line, NS0-derived recombinant human Klotho beta with a C-terminal 10 His tag. Phe53-Leu997 Accession # Q86Z14 | |
RUO | |
152831 | |
Human | |
IgG |
Flow Cytometry | |
1025C | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q86Z14 | |
Rabbit | |
100 μg | |
Primary | |
Detects human Klotho beta in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction