Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Klotho beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
| Antigen | Klotho beta |
|---|---|
| Clone | 885935 |
| Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Klotho beta Monoclonal specifically detects Klotho beta in Human samples. It is validated for Western Blot, Simple Western.Specifications
| Klotho beta | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| KLB | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 885935 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q86Z14 | |
| 152831 | |
| Mouse myeloma cell line NS0-derived recombinant human Klotho beta Phe53-Leu997 Accession # Q86Z14 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Klotho beta in direct ELISA and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title