Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Klotho beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF5889
This item is not returnable.
View return policy
Description
Klotho beta Polyclonal specifically detects Klotho beta in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Klotho beta | |
Polyclonal | |
Unconjugated | |
Q86Z14 | |
KLB | |
Mouse myeloma cell line NS0-derived recombinant human Klotho beta Phe53-Leu997 Accession # Q86Z14 | |
100 μg | |
Primary | |
Detects human Klotho beta in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant mouse Klotho beta is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
Goat | |
Affinity Purified | |
RUO | |
152831 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction