Learn More
Gibco™ Human Klotho Recombinant Protein, PeproTech®

Description
100-53-1MG will be provided as 2 x 500 μg (100-53-500UG). Recombinant Human Klotho is a glycoprotein of 516 amino acid residues that migrates at an apparent molecular weight of 65-70 kDa by SDS-PAGE analysis under reducing conditions. Recombinant Human Klotho has a calculated molecular weight of 58.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Specifications
Specifications
Accession Number | Q9UEF7 |
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 9365 |
Molecular Weight (g/mol) | 58.6 kDa |
Name | Human Klotho |
Quantity | 5 μg |
Source | CHO cells |
Regulatory Status | RUO |
Endotoxin Concentration | <1 EU/ μg |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.