Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KLRG1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$212.05 - $555.75
Specifications
| Antigen | KLRG1 |
|---|---|
| Clone | 2388C |
| Dilution | Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB70293SP
![]() |
R&D Systems
MAB70293SP |
25 μg |
Each for $212.05
|
|
|||||
MAB70293100
![]() |
R&D Systems
MAB70293100 |
100 μg |
Each for $555.75
|
|
|||||
Description
KLRG1 Monoclonal specifically detects KLRG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| KLRG1 | |
| Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q96E93 | |
| 10219.0 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2388C | |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
| Chinese hamster ovary cell line CHO-derived recombinant human KLRG1, Leu60-Phe195, Accession No. Q96E93 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human KLRG1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title