Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KLRG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB70291
This item is not returnable.
View return policy
Description
KLRG1 Monoclonal specifically detects KLRG1 in Human samples. It is validated for Western Blot.Specifications
KLRG1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q96E93 | |
KLRG1 | |
Chinese hamster ovary cell line CHO-derived recombinant human KLRG1 Leu60-Phe195 Accession # Q96E93 | |
100 μg | |
Primary | |
Detects human KLRG1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
697213 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10219 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction