Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KOR Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3895S100UG
This item is not returnable.
View return policy
Description
KOR Monoclonal specifically detects KOR in Human samples. It is validated for Flow Cytometry.Specifications
KOR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P41145 | |
OPRK1 | |
NS0 mouse myeloma cell line transfected with human KOR Met1-Val380 Accession # P41145 | |
RUO | |
4986 | |
Human | |
IgG1 |
Flow Cytometry | |
387301 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK, OPRK1, Oprk2, R21 | |
Mouse | |
100 μg | |
Primary | |
Detects human KOR. Stains human KOR transfectants but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction