Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KOR Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $556.04
Specifications
| Antigen | KOR |
|---|---|
| Clone | 387301 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
Description
KOR Monoclonal specifically detects KOR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| KOR | |
| LYOPH | |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P41145 | |
| 4986 | |
| NS0 mouse myeloma cell line transfected with human KOR Met1-Val380 Accession # P41145 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human KOR. Stains human KOR transfectants but not irrelevant transfectants. |
| 387301 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK, OPRK1, Oprk2, R21 | |
| OPRK1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title