Learn More
Human Kremen-1 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Supplier:  R&D Systems AF2127
Catalog No. AF2127
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Kremen-1 Polyclonal specifically detects Kremen-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Kremen-1
Polyclonal
Unconjugated
NP_114434
KREMEN1
Mouse myeloma cell line NS0-derived recombinant mouse Kremen-1 Ala20-Gly397 Accession # NP_114434
100 μg
Primary
Detects human Kremen-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse Kremen-1 and less than 5% cross-reactivity with recombinant human Kremen-2 is observed.
Human
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, Kremen1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1, KRM1Dickkopf receptor
Goat
Affinity Purified
RUO
83999
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.