Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Kynureninase Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$214.00 - $538.00

Specifications

Antigen Kynureninase
Clone 589731R
Dilution Western Blot 2 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB4887RSP
GREENER_CHOICE
View Documents
R&D Systems
MAB4887RSP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB4887R100
GREENER_CHOICE
View Documents
R&D Systems
MAB4887R100
100 μg
Each for $538.00
Only null left
Add to Cart
 
Description

Description

Kynureninase Monoclonal specifically detects Kynureninase in Human samples. It is validated for Western Blot.
Specifications

Specifications

Kynureninase
Western Blot 2 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
EC 3.7.1.3, KYNU, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Kynureninase, Met1-Asn465, Accession # Q16719
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Kynureninase in direct ELISAs.
589731R
Western Blot
Unconjugated
Mouse
Human
Q16719
8942
IgG1
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.