Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human L-Selectin/CD62L Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000099
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000099 Supplier R&D Systems Supplier No. FAB9787V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Human samples. It is validated for Flow Cytometry

Specifications

Antigen L-Selectin/CD62L
Applications Flow Cytometry
Classification Monoclonal
Clone 4G8
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1
Host Species Mouse
Immunogen Recombinant human L-Selectin/CD62L, Extracellular domain
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6402
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.