Learn More
Human L-Selectin/CD62L Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | L-Selectin/CD62L |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4G8 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1 |
| Host Species | Mouse |
| Immunogen | Recombinant human L-Selectin/CD62L Extracellular domain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.