Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human L1CAM Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7771T100UG
This item is not returnable.
View return policy
Description
L1CAM Monoclonal specifically detects L1CAM in Human samples. It is validated for Flow Cytometry.Specifications
L1CAM | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
antigen identified by monoclonal R1, CAML1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Homogenous suspension of 16 week human fetal brain., Accession # P32004 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
UJ127 | |
Flow Cytometry 0.25-1 ug/106 cells | |
P32004 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
3897 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction