Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human L1CAM ELISA Kit (Colorimetric)
SDP

Catalog No. NB230266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB230266 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB230266 Supplier Novus Biologicals Supplier No. NBP280312

Specifications

Assay Range 39.06-2500 pg/mL
Assay Sensitivity 8.31 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity Recognizes both recombinant and natural Human L1CAM.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3897
Gene Symbol L1CAM
Quantity 1 Kit
Research Discipline Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
Storage Requirements Storage is content dependent.
Synonym antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.