Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lactate Dehydrogenase B/LDHB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9205100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9205100 100 μg
MAB9205SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9205100 Supplier R&D Systems Supplier No. MAB9205100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lactate Dehydrogenase B Monoclonal specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated.

Specifications

Antigen Lactate Dehydrogenase B
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 2057D
Conjugate Unconjugated
Dilution Western Blot 0.05 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P07195
Gene Alias EC 1.1.1, EC 1.1.1.27, Epididymis Secretory Protein Li 281, HEL-S-281, lactate dehydrogenase B, LDH heart subunit, LDHB, LDH-B, LDHBD, LDH-H, L-lactate dehydrogenase B chain, Renal Carcinoma Antigen NY-REN-46, TRG-5
Host Species Rabbit
Immunogen E. coli-derived recombinant human LDHB Met1-Val93 Accession # P07195
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3945
Test Specificity Detects human LDHB in direct ELISAs and Western blots.
Reconstitution Reconstitue at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.