Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LAG-3 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB23194G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB23194G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB23194G100 Supplier R&D Systems Supplier No. FAB23194G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LAG-3
Applications Flow Cytometry
Classification Monoclonal
Clone 1009614
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P18627
Gene Alias CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAG-3, Met1-Leu450, Accession # P18627
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3902
Test Specificity Detects human LAG-3 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.