Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LAG-3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB23194T100UG
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry.Specifications
LAG-3 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
Mouse myeloma cell line NS0-derived recombinant human LAG-3, Met1-Leu450, Accession # P18627 | |
100 μg | |
3902 | |
Human | |
Supernatant |
Flow Cytometry | |
1009614 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P18627 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human LAG-3 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction