Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LAG-3 (CD223) Recombinant Protein, Biotin

Catalog No. PIRP88097
Encompass
Click to view available options
Quantity:
100 μg
20 μg

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18627
Concentration 0.25 mg/mL
For Use With (Application) ELISA
Format Lyophilized
Formulation PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 3902
Molecular Weight (g/mol) 47.0 kDa
Name Human LAG-3 (CD223)
Purification Method SDS-PAGE
Quantity 20 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/ μg
Gene Alias Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3 lymphocyte activating 3; soluble LAG3
Common Name LAG-3 (CD223)
Gene Symbol LAG3
Biological Activity Immobilized Human LAG3, Biotinylated (at 2 μg/mL (100 μL/well), EC50 is 5-15 ng/mL Can bind Human FGL1 (hFc Tag), the EC50 is 20-65 ng/mL)
Product Type Protein
Conjugate Biotin
Recombinant Recombinant
Sequence Human LAG3, amino acids Met1-Arg440 (Accession # NP_002277.4)
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized Human LAG3, Biotinylated (at 2 μg/mL (100 μL/well) can bind Human FGL1 (hFc Tag), the EC50 is 150-700 ng/mL)
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.