Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LAIR1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2664G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2664G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2664G100 Supplier R&D Systems Supplier No. FAB2664G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LAIR1 Monoclonal specifically detects LAIR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LAIR1
Applications Flow Cytometry
Classification Monoclonal
Clone 342219
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q6GTX8
Gene Alias CD305, CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1
Gene Symbols LAIR1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1 Gln22-His163 Accession # Q6GTX8
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3903
Test Specificity Detects human LAIR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LAIR2 or recombinant mouse LAIR1 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.