Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LAIR1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | LAIR1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
LAIR1 Polyclonal specifically detects LAIR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
LAIR1 | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD305, CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
LAIR1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q6GTX8 | |
3903 | |
Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1 Gln22-His163 Accession # Q6GTX8 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human LAIR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human LAIR2 is observed and less than 2% cross-reactivity with recombinant mouse LAIR1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title