Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LAIR2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2665U100UG
This item is not returnable.
View return policy
Description
LAIR2 Monoclonal specifically detects LAIR2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
LAIR2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6ISS4 | |
LAIR2 | |
Mouse myeloma cell line NS0-derived recombinant human LAIR2 isoform 1 Gln22-Pro152 Accession # Q6ISS4 | |
RUO | |
3904 | |
Human | |
IgG2b |
Flow Cytometry | |
319701 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD306, CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634 | |
Mouse | |
100 μg | |
Primary | |
Detects human LAIR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LAIR1, recombinant mouse (rm) LAIR1 or rmLAIR2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction