Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Lambda Light Chain ELISA Kit (Colorimetric)
SDP

Catalog No. NB600831KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600831KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600831KT Supplier Novus Biologicals™ Supplier No. NBP2600831KIT
Add to Cart
Add to Cart

Colorimetric ELISA Kit is useful in the study of immunological pathways

The Human Lambda Light Chain ELISA Kit (Colorimetric) measures human Lambda Light Chain in serum, plasma.

Specifications

Assay Range 0.039 to 10μg/mL
Assay Sensitivity 0.021μg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3536
Gene Symbol IGL, IGLC2
Kit Contents Assay plate, Standard (Freeze dried), HRP-conjugate (100X concentrate), HRP- conjugate Diluents, Sample Diluents, Wash Buffer (25X concentrate), TMB
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Immunology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym IGLC, immunoglobulin lambda constant group
Target Lambda Light Chain
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.