Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lamin B1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$212.05 - $473.10
Specifications
| Antigen | Lamin B1 |
|---|---|
| Clone | 919007 |
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8525SP
![]() |
R&D Systems
MAB8525SP |
25 μg |
Each for $212.05
|
|
|||||
MAB8525100
![]() |
R&D Systems
MAB8525100 |
100 μg |
Each for $473.10
|
|
|||||
Description
Lamin B1 Monoclonal specifically detects Lamin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Lamin B1 | |
| Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P20700 | |
| 4001 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 919007 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, LMNB1, MGC111419 | |
| E. coli-derived recombinant human Lamin B1 Arg418-Ser582 Accession # P20700 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Lamin B1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title