Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Laminin alpha 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2143100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2143100 100 μg
MAB2143SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2143100 Supplier R&D Systems Supplier No. MAB2143100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Laminin alpha 2 Monoclonal specifically detects Laminin alpha 2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Laminin alpha 2
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1003531
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P24043
Gene Alias LAMA2, Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMM, LAMMLaminin-4 subunit alpha, Merosin, Merosin heavy chain
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Laminin alpha 2, Lys2145-Asn3122, Accession # P24043
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3908
Test Specificity Detects human Laminin alpha 2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.