Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Laminin alpha 4 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF7340SP
This item is not returnable.
View return policy
Description
Laminin alpha 4 Polyclonal specifically detects Laminin alpha 4 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization, Immunocytochemistry.Specifications
Laminin alpha 4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp686D23145, LAMA3LAMA4*-1, LAMA4, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha | |
Sheep | |
Affinity Purified | |
RUO | |
3910 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
LYOPH | |
Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Neutralization 1.5-7.5 μg/mL, Immunocytochemistry 5-15 μg/mL | |
EAW48267 | |
LAMA4 | |
Chinese hamster ovary cell line CHO-derived recombinant human Laminin alpha 4 Gln826-Ala1816, predicted Accession # EAW48267 | |
25 μg | |
Primary | |
Detects human Laminin alpha 4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Laminin alpha 4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction