Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LAMP-1/CD107a Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4800G
This item is not returnable.
View return policy
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human samples. It is validated for Flow Cytometry.Specifications
LAMP-1/CD107a | |
Monoclonal | |
Alexa Fluor 488 | |
P11279 | |
LAMP1 | |
Mouse myeloma cell line NS0-derived recombinant human LAMP-1/CD107a Ala28-Asn380 Accession # P11279 | |
100 Tests | |
3916 | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry | |
508921 | |
Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 1:10 dilution | |
CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP1, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human LAMP1/CD107a in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction