Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LAMP-2/CD107b Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6228V100UG
This item is not returnable.
View return policy
Description
LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
LAMP-2/CD107b | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P13473 | |
LAMP2 | |
Mouse myeloma cell line NS0-derived recombinant human LAMP2/CD107b Leu29-Phe375 Accession # P13473 | |
RUO | |
3920 | |
Human | |
IgG1 |
Flow Cytometry | |
743320 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP2, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane p | |
Mouse | |
100 μg | |
Primary | |
Detects human LAMP2/CD107b in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LAMP1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction